Labconco™ Axiom™ Class II, Type C1 Biosafety Cabinet
Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination Technology and MyLogic Operating System.
Supplier:  Labconco™ 304611100
 View more versions of this product
Catalog No. 30-461-1100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
  • Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
  • Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes
  • C1 designation is pending NSF approval
Specifications
Specifications
NSF International-Listed, ETL, Modified ASHRAE 110
North America
25.4 cm
UV Light, Service Fixture, Vacu-Pass Portal, Base Stand
245.1 cm
78.2 in.
32.6 in.
1000 lb.
115 V, 60 Hz, 12 A, Domestic
Safety Cabinet
96.5 in.
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Fluorescent, UV
10 in.
198.6 cm
82.8 cm
453.6 kg