Purgeable Mixtures (For EPA 624)
$127.37 - $127.37
Chemical Identifiers
Chemical Name or Material Purgeable Mixtures
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-PMX-1101 Ultra Scientific
PMX1101
Ampule 1x1mL, 100μg/mL Each for $127.37
Specifications Description & Specifications
Chemical Identifiers
Purgeable Mixtures
Specifications
1x1mL, 100μg/mL
Each component at 20μg/mL (US-PMX-100) or 100μg/mL (US-PMX-110) in Methanol:
Benzene
Bromodichloromethane
Bromoform
Bromomethane
Carbon Tetrachloride
Chlorobenzene
Chloroethane
Chloroform
Chloromethane
Dibromochloromethane
1,2-Dichlorobenzene
1,3-Dichlorobenzene
1,4-Dichlorobenzene
1,1-Dichloroethane
1,2-Dichloroethane
1,1-Dichloroethene
trans-1,2-Dichloroethene
1,2-Dichloropropane
cis-1,3-Dichloropropene
trans-1,3-Dichloropropene
Ethylbenzene
Methylene Chloride
1,1,2,2-Tetrachloroethane
Tetrachloroethene
Toluene
1,1,1-Trichloroethane
1,1,2-Trichloroethane
Trichloroethene
Trichlorofluoromethane
Vinyl Chloride
Purgeable Mixtures
For EPA Method 624
Ampule