missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Purell™ Hand Sanitizing Wipes High Capacity Floor Stand Dispenser

High capacity floor stand dispenser for PURELL™ wipes

$306.15 - $551.41

Specifications

Product Type Stand Dispenser
Capacity 6000 Wipes
For Use With (Equipment) PURELL™ Hand Sanitizing Wipes
Quantity 1 Ea.
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
19-043-941
View Documents Promotion Details
GOJO Canada Inc
9118DS2B
Black
Case for $551.41
 
19-043-942
View Documents Promotion Details
GOJO Canada Inc
9118DSLV
Silver
Case for $306.15
 
Description

Description

  • Weighted base keeps stand in place
  • Durable, freestanding unit
  • Pre-installed SIMPUL™ nozzle technology, virtually eliminates roping, providing one wipe every time
  • Storage compartment fits 6,000 wipes
  • Available with customizable signage
  • Available in blue, black and reflective silver
Specifications

Specifications

Stand Dispenser
6000 Wipes
PURELL™ Hand Sanitizing Wipes
1 Ea.
SDS