Learn More
BIOTIX™ Pure uTIP™ Filter Sterile Universal Pipette Tips
Pure uTIP™ Filter Pipette Tips for Universal Pipettes, Standard Designed with Flexfit™ to lower necessary insertion and ejection force by ∼70% to make secure seal. Manufactured with Pure resin that is virgin polypropylene.
$186.90 - $278.57
Specifications
| Sterility | Sterile |
|---|---|
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Format | Hinged Rack Sterile |
| Product Type | Pipette Tips |
| Catalog Number | Mfr. No. | Volume (Metric) | Tip Style | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Tip Style | Price | Quantity | ||||
|
12-111-171
|
BIOTIX™
63305403 |
2 μL | Ergonomic, Finely Tapered, Fine Point, Flextop, Non-beveled Point |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-170
|
BIOTIX™
63305404 |
10 μL | Fine Point, Finely Tapered, Flextop, Ergonomic |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-169
|
BIOTIX™
63305405 |
10 μL | Extended Length, Fine Point, Finely Tapered, Ergonomic |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-141
|
BIOTIX™
63305451 |
10 μL | Ergonomic, Extended Length, Fine Point, Finely Tapered, Non-beveled Point, Eppendorf Style |
Pack of 960 Each for $186.90
Case of 5 Pack for $934.50
|
|
||||
|
12-111-168
|
BIOTIX™
63305406 |
20 μL | Fine Point, Finely Tapered, Ergonomic |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-167
|
BIOTIX™
63305407 |
100 μL | Ergonomic, Flextop, Fine Point, Finely Tapered, Non-beveled Point |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-166
|
BIOTIX™
63305408 |
200 μL | Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-165
|
BIOTIX™
63305409 |
300 μL | Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point |
Pack of 960 Each for $222.87
Case of 5 Pack for $1,114.35
|
|
||||
|
12-111-164
|
BIOTIX™
63305410 |
1000 μL | Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point |
Pack of 960 Each for $278.57
Case of 4 Pack for $1,114.28
|
|
||||
|
12-111-163
|
BIOTIX™
63305411 |
1250 μL | Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point |
Pack of 960 Each for $278.57
Case of 4 Pack for $1,114.28
|
|
||||
Description
Biotix™ Pure uTIP™ Filter Pipette Tips for Universal Pipettes, Standard Designed with Flexfit™ to lower necessary insertion and ejection force by about 70% to make secure seal. Biotix™ uTIP™ Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin that is virgin polypropylene.
- Fits all standard pipettes
- FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
Specifications
| Sterile | |
| Not Autoclavable | |
| Pipette Tips | |
| Yes |
| Clear | |
| Hinged Rack Sterile | |
| Single-use | |
| uTip |
This product contains no safety hazards