Please login to your online account to display your discounted pricing
Whatman™ Puradisc Syringe Filters, Nonsterile: PROMO OFFER
Low hold-up volume—less than 0.1mL with air purge
$591.00 - $591.00
Specifications
Type Syringe Filter
Inlet Style female Luer-Lok
View More Specs
Products
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity    
05713399PM
whatman™
67852502PM
Polytetrafluoroethylene (PTFE) 0.2μm 200/pk. Pack of 200 Each for $591.00
Description & Specifications
Specifications
Type Syringe Filter
Inlet Style female Luer-Lok
Disposable syringe filters for purifying aqueous and organic solutions. All six filter types have polypropylene housings, female Luer-Lock™ inlet and male slip Luer outlet.
Puradisc PES
  • Polyethersulfone (PES) membrane for filtering aqueous solutions
  • Low protein binding
  • Nonsterile packages or radiation-sterilized blister packs
Puradisc GD
  • Graded-density membrane, very high loading capacity
  • Designed for HPLC, TLC or GC applications requiring 1μm particle retention
  • Use for filtering high debris, hard-to-filter solutions
  • Made of multilayered glass microfiber
  • Nonsterile
Puradisc GF/F
  • High loading capacity for air/gas filtration, HPLC, TLC, and GC sampling
  • Membrane made of binder-free glass microfiber
  • Nonsterile
Puradisc NYL
  • For filtering aqueous or solvent-based solutions, HPLC, TLC, and GC sampling
  • Membrane made of nylon
  • Nonsterile
Puradisc TF
  • For filtering solvents, chemicals and nonaqueous solutions
  • Typical applications include HPLC, TLC, and GC sample clarification; HPLC solvent filtration
  • Made of hydrophobic polytetrafluoroethylene (PTFE)
  • Nonsterile, autoclavable
Puradisc PP
  • Polypropylene media designed for aqueous and organic solutions containing high debris levels and for hard-to-filter solutions
  • Nonsterile, autoclavable