missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-Pulegone, TCI America™

Manufacturer:  TCI America P233125ML

 View more versions of this product

Catalog No. P233125ML



Specifications

Specifications

(+)-Pulegone
≥85.0% (GC)
MFCD00063000
Yellow
(+)-1-Methyl-4-isopropylidene-3-cyclohexanone
CC1CCC(=C(C)C)C(=O)C1
152.237
CHEBI:81226
Clear Liquid at 20°C
89-82-7
C10H16O
217°C
25mL
NZGWDASTMWDZIW-QMMMGPOBSA-N
(5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
638012
152.24

Chemical Identifiers

89-82-7
152.237
NZGWDASTMWDZIW-QMMMGPOBSA-N
638012
C10H16O
MFCD00063000
(+)-1-Methyl-4-isopropylidene-3-cyclohexanone
CHEBI:81226
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2400

RTECSNumber : OT0261000

TSCA : Yes

Documents