Please login to your online account to display your discounted pricing

Pulegone, 92%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC418505000

 View more versions of this product

Catalog No. AC418505000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥90%
Physical Form Liquid
Formula Weight 152.24
Purity Grade Practical Grade
Quantity 500g
Color Clear colorless to yellow
Density 0.931g/mL
Specific Rotation Condition +21° (20°C neat)
Merck Index 15, 8046
Beilstein 07, 81
Packaging Glass bottle
CAS 89-82-7
Chemical Name or Material Pulegone
Molecular Formula C10H16O
Assay Percent Range 92%
MDL Number MFCD00063000
Refractive Index 1.4850 to 1.4880 (20°C, 589nm)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Pulegone
CAS 89-82-7
Purity Grade Practical Grade
Assay ≥90%
Assay Percent Range 92%
Molecular Formula C10H16O
Formula Weight 152.24
MDL Number MFCD00063000