Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's G Balanced Salt Solution, without calcium, without magnesium, without phenol red

Chemical Identifiers

Chemical Name or Material Puck's G Balanced Salt Solution
Name Note without calcium, without magnesium, without phenol red
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67516AP

 
alfa aesar™
J67516AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67516K2

 
alfa aesar™
J67516K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Puck's G Balanced Salt Solution
Physical Form Liquid
TSCA TSCA
Color Colorless
Quantity 500mL
Name Note without calcium, without magnesium, without phenol red