Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's G Balanced Salt Solution, without calcium, without magnesium, without phenol red


Quantity 1L

Manufacturer: alfa aesar™  J67516K2

 View more versions of this product

Catalog No. AAJ67516K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Puck's G Balanced Salt Solution
Physical Form Liquid
Name Note without calcium, without magnesium, without phenol red
TSCA TSCA
Color Colorless
Quantity 1L

Specifications

Chemical Name or Material Puck's G Balanced Salt Solution
Name Note without calcium, without magnesium, without phenol red