Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's G Balanced Salt Solution, with calcium, with magnesium, with phenol red


Quantity 500mL

Manufacturer: alfa aesar™  J67542AP

 View more versions of this product

Catalog No. AAJ67542AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Puck's G Balanced Salt Solution
TSCA TSCA
Name Note with calcium, with magnesium, with phenol red
Physical Form Liquid
Color Red
Quantity 500mL

Specifications

Chemical Name or Material Puck's G Balanced Salt Solution
Name Note with calcium, with magnesium, with phenol red