Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's F Balanced Salt Solution (1X), with calcium, with magnesium, with phenol red


Quantity 1L

Manufacturer: alfa aesar™  J67665K2

 View more versions of this product

Catalog No. AAJ67665K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Puck's F Balanced Salt Solution (1X)
Physical Form Liquid
Name Note with calcium, with magnesium, with phenol red
TSCA TSCA
Color Red
Quantity 1L

Specifications

Chemical Name or Material Puck's F Balanced Salt Solution (1X)
Name Note with calcium, with magnesium, with phenol red