Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's F Balanced Salt Solution (10X), without calcium, without magnesium, without phenol red


Quantity 500mL

Manufacturer: alfa aesar™  J67654AP

 View more versions of this product

Catalog No. AAJ67654AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

TSCA TSCA
Name Note without calcium, without magnesium, without phenol red
Color Colorless
Quantity 500mL
Physical Form Liquid
Chemical Name or Material Puck's F Balanced Salt Solution (10X)

Specifications

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, without phenol red