Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's F Balanced Salt Solution (10X), without calcium, without magnesium, without phenol red

Chemical Identifiers

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, without phenol red
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67654AP

  SDS
alfa aesar™
J67654AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67654K2

  SDS
alfa aesar™
J67654K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Name Note without calcium, without magnesium, without phenol red
Color Colorless
Quantity 500mL
Physical Form Liquid
Chemical Name or Material Puck's F Balanced Salt Solution (10X)