Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's F Balanced Salt Solution (10X), without calcium, without magnesium, with phenol red


Quantity 500mL

Manufacturer: alfa aesar™  J67781AP

 View more versions of this product

Catalog No. AAJ67781AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
Physical Form Liquid
Name Note without calcium, without magnesium, with phenol red
Quantity 500mL
TSCA TSCA
Color Red

Specifications

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, with phenol red