Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's F Balanced Salt Solution (10X), without calcium, without magnesium, with phenol red

Chemical Identifiers

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, with phenol red
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67781AP

 
alfa aesar™
J67781AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67781K2

 
alfa aesar™
J67781K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
TSCA TSCA
Color Red
Quantity 500mL
Name Note without calcium, without magnesium, with phenol red
Physical Form Liquid