Please login to your online account to display your discounted pricing

Alfa Aesar™ Puck's F Balanced Salt Solution (10X), without calcium, without magnesium, with phenol red


Quantity 1L

Manufacturer: alfa aesar™  J67781K2

 View more versions of this product

Catalog No. AAJ67781K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
TSCA TSCA
Color Red
Quantity 1L
Name Note without calcium, without magnesium, with phenol red
Physical Form Liquid

Specifications

Chemical Name or Material Puck's F Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, with phenol red