Please login to your online account to display your discounted pricing

PU 02, Tocris™

Negative allosteric modulator of 5-HT3

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material PU 02
Assay Percent Range >97%
Molecular Formula C16H12N4S
Formula Weight 292.36
Synonym 6-[(1-Naphthalenylmethyl)thio]-9H-purine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

470050

 
r & d systems
470050
50mg Each for $746.48

470010

 
r & d systems
470010
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 292.36
Quantity 50mg
Synonym 6-[(1-Naphthalenylmethyl)thio]-9H-purine
CAS 313984-77-9
Chemical Name or Material PU 02
Assay Percent Range >97%
Molecular Formula C16H12N4S
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C