Please login to your online account to display your discounted pricing

PTP1B Inhibitor

$337.11 - $337.11

Specifications

CAS 765317-72-4
Formula Weight 741.45g/mol
Synonym 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethyl-N-[4-(2-thiazolylsulfamoyl)phenyl]benzofuran-6-sulfonamide
Storage Requirements −30° to −10°C
Format Solid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64512LB0

 
alfa aesar™
J64512LB0
Each for $337.11
Description & Specifications

Specifications

CAS 765317-72-4
Formula Weight 741.45g/mol
Synonym 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethyl-N-[4-(2-thiazolylsulfamoyl)phenyl]benzofuran-6-sulfonamide
Storage Requirements −30° to −10°C
Format Solid
Sensitivity Light Sensitive
For Use With (Application) PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway
Description White
Solubility Soluble in ethanol and DMSO

  • PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway