PTK6 (BRK) Recombinant Human Protein DFS Item
PTK6 (BRK) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3291
Catalog No. PV3291
Description
Description
Breast tumor kinase (Brk) is a non-receptor protein tyrosine kinase expressed in over 60% of breast carcinoma tissue samples and breast tumor cell lines.
Specifications
Specifications
NP_005966
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
55.4kDa
95% by SDS-PAGE
–80°C
1 Each
PTK6 (Brk)
PTK6 (Brk) was pre-diluted in enzyme dilution buffer and assayed in 10mM HEPES (pH 7.5), 150mM NaCl, 10 MnCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
SRC
190nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
0.27mg/mL
5753
PTK6 (BRK) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
PTK6 (BRK) Recombinant Human Protein
Kinase assay
Baculovirus
PTK6 (BRK) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.