Invitrogen™ PTK2B (FAK2) Recombinant Human Protein DFS Item
PTK2B (FAK2) is a cytoplasmic tyrosine kinase.
Manufacturer:  Invitrogen™ LSPV4567
 View more versions of this product
Catalog No. PV4567
Description
Description
PTK2B (FAK2) is involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway.
Specifications
Specifications
NP_004094
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
10μg
RUO - research use only
Full Length, Recombinant, Cytoplasmic
PTK2B (FAK2)
Kinase Assay
Baculovirus
Human
GST Tag
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
145.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
N-Terminal
Cytoplasmic Tyrosine Kinases
PTK2B protein tyrosine kinase 2 beta
Kinases & Inhibitors
FAK Kinase Family
Yes
Documents