Invitrogen™ PTK2 (FAK) Recombinant Human Protein DFS Item
PTK2 (FAK) is a cytoplasmic protein tyrosine kinase.
Manufacturer:  Invitrogen™ LSPV4085
 View more versions of this product
Catalog No. PV4085
Description
Description
PTK2 (FAK) is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents.
Specifications
Specifications
NP_722560
Frozen
146.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
FAK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PTK2 (FAK), FADK, FAK1
Baculovirus
Human
GST Tag
Documents