Learn More
Sartorius PTFE (Polytetrafluorethylene) Membrane Filters
Filter air and gasses, even at low differential pressures. Sartorius PTFE (Polytetrafluorethylene) Membrane Filters are permanently hydrophobic with low adsorption and excellent chemical compatibility. Choose from four pore sizes in a variety of diameters.
$410.34 - $732.02
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Color | White |
| Disposable | Single-use |
| Filter Type | Membrane Filter |
| Material | PTFE |
| Catalog Number | Mfr. No. | Diameter (Metric) | Pore Size | Thickness (Metric) | Bubble Point | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Pore Size | Thickness (Metric) | Bubble Point | Price | Quantity | ||
|
14-555-415
|
Sartorius
11806100G |
100 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 25 Each for $670.17
|
|
||
|
14-555-423
|
Sartorius
1180713N |
13 mm | 0.2 μm | 65 μm | 1.4bar |
Pack of 100 Each for $410.34
|
|
||
|
14-555-422
|
Sartorius
11807100G |
100 mm | 0.2 μm | 65 μm | 1.4bar |
Pack of 25 Each for $670.17
|
|
||
|
14-555-418
|
Sartorius
1180637N |
37 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 100 Each for $670.17
|
|
||
|
14-555-417
|
Sartorius
1180625N |
25 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 100 Each for $490.76
|
|
||
|
14-555-430
|
Sartorius
1184250N |
50 mm | 5 μm | 100 μm | 0.1bar |
Pack of 100 Each for $732.02
|
|
||
|
14-555-419
|
Sartorius
1180647N |
47 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 100 Each for $670.17
|
|
||
|
14-555-424
|
Sartorius
1180725N |
25 mm | 0.2 μm | 65 μm | 1.4bar |
Pack of 100 Each for $490.76
|
|
||
|
14-555-416
|
Sartorius
1180613N |
13 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 100 Each for $410.34
|
|
||
|
14-555-412
|
Sartorius
1180347N |
47 mm | 1.2 μm | 100 μm | 0.45bar |
Pack of 100 Each for $732.02
|
|
||
|
14-555-414
|
Sartorius
1180390G |
90 mm | 1.2 μm | 100 μm | 0.45bar |
Pack of 25 Each for $590.78
|
|
||
|
14-555-427
|
Sartorius
11842100G |
100 mm | 5 μm | 100 μm | 0.1bar |
Pack of 25 Each for $732.02
|
|
||
|
14-555-429
|
Sartorius
1184247N |
47 mm | 5 μm | 100 μm | 0.1bar |
Pack of 100 Each for $732.02
|
|
||
|
14-555-428
|
Sartorius
1184225N |
25 mm | 5 μm | 100 μm | 0.1bar |
Pack of 100 Each for $582.53
|
|
||
|
14-555-413
|
Sartorius
1180350N |
50 mm | 1.2 μm | 100 μm | 0.45bar |
Pack of 100 Each for $732.02
|
|
||
|
14-555-425
|
Sartorius
1180747N |
47 mm | 0.2 μm | 65 μm | 1.4bar |
Pack of 100 Each for $670.17
|
|
||
|
14-555-421
|
Sartorius
1180690G |
90 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 25 Each for $670.17
|
|
||
|
14-555-411
|
Sartorius
1180325N |
25 mm | 1.2 μm | 100 μm | 0.45bar |
Pack of 100 Each for $582.53
|
|
||
|
14-555-426
|
Sartorius
1180790G |
90 mm | 0.2 μm | 65 μm | 1.4bar |
Pack of 25 Each for $670.17
|
|
||
|
14-555-420
|
Sartorius
1180650N |
50 mm | 0.45 μm | 80 μm | 0.94bar |
Pack of 100 Each for $670.17
|
|
||
Description
Filter air and gasses, even at low differential pressures. Sartorius PTFE (Polytetrafluorethylene) Membrane Filters are permanently hydrophobic with low adsorption and excellent chemical compatibility. Choose from four pore sizes in a variety of diameters.
Constructed of pure PTFE to be permanently hydrophobic
- Not wetted by air humidity to allow unhindered passage of air
- Excellent chemical compatibility (pH 1 to 14) and low adsorption (Gamma Globulin 8 μg/cm²
- Lab applications:
- The main application is air/gas filtration, even at low differential pressures
- Also ideal for filtration of solvents and acids to which other filter types are not resistant
(Must be prewetted with ethanol or methanol before filtration of aqueous media)
- A variety of non-sterile, white membranes without grids:
- 0.2 μm, 0.45 μm, 1.2 μm and 5.0 μm pore sizes
- 13, 25, 37, 47, 50, 90 and 100 mm diameters
- Can be sterilized by autoclaving at 121° or 134°C or with ethylene oxide
- Bulk packed in boxes of 100 (boxes of 25 for the 90 or 10 0 mm filters)
Specifications
| Autoclavable | |
| Single-use | |
| PTFE | |
| Air/gas-filtration | |
| Membrane Filter |
| White | |
| Membrane Filter | |
| PTFE | |
| Hydrophobic |