missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PTEN/MMAC1 Ab-4, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ PTEN/MMAC1 Ab-4, Mouse Monoclonal Antibody.

$374.02 - $1,748.51

Specifications

Antigen PTEN/MMAC1 Ab-4
Clone 17.A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS1601S View Documents Thermo Scientific™
MS1601S
1mL Each for $1,748.51
 
MS1601S0 View Documents Thermo Scientific™
MS1601S0
100μL Each for $374.02
 
MS1601S1 View Documents Thermo Scientific™
MS1601S1
500μL Each for $1,190.24
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ PTEN/MMAC1 Ab-4, Mouse Monoclonal Antibody.

PTEN/MMAC1, a tumor suppressor, has been implicated in a large number of human tumors and is conserved from humans to worms. Characterization of PTEN protein showed that it is a phosphatase that acts on proteins and on 3-phosphorylated phosphoinositides, and can therefore modulate signal-transduction pathways that involve lipid second messengers. Recent results indicate that at least part of its role is to regulate the activity of the serine/threonine kinase AKT/PKB, and thus influence cell survival signaling.

Host Species: Mouse

Clone: 17.A

Isotype: IgM/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human PTEN protein

Molecular Weight: 60kDa

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

PTEN/MMAC1 Ab-4
Immunohistochemistry (Paraffin)
Unconjugated
Murine
Cancer and Tumor Biology
Recombinant human PTEN protein
17.A
Monoclonal
Concentrated
RUO
Primary
IgM kappa