Please login to your online account to display your discounted pricing
Thermo Scientific™ T7 Cell-Free Expression Vectors
Generate His-, HA-, Myc- or other label-tagged fusion proteins by cell-free in vitro translation (IVT) with these versatile T7 expression vectors.
$130.91 - $159.79
Products
Catalog Number Mfr. No. Description Price Quantity    
PI88859
thermo scientific™
88859
pT7CFE1-NHis Vector Each for $159.38
PI88860
thermo scientific™
88860
pT7CFE1-CHis Vector Each for $130.91
PI88861
thermo scientific™
88861
pT7CFE1-NHA Vector Each for $159.79
PI88862
thermo scientific™
88862
pT7CFE1-CHA Each for $159.79
PI88863
thermo scientific™
88863
pT7CFE1-NMyc Vector Each for $159.79
PI88864
thermo scientific™
88864
pT7CFE1-CMyc Vector Each for $159.79
PI88865
thermo scientific™
88865
pT7CFE1-NFtag Vector Each for $159.38
PI88866
thermo scientific™
88866
pT7CFE1-CFtag Vector Each for $159.38
PI88867
thermo scientific™
88867
pT7CFE1-NHA-CHis Vector Each for $159.21
PI88868
thermo scientific™
88868
pT7CFE1-CGST-HA-His Vector Each for $159.27
PI88869
thermo scientific™
88869
pT7CFF1-CGFP-HA-His Vector Each for $159.38
Description & Specifications
Thermo Scientific™ T7 Cell-Free Expression Vectors (pT7CFE1) are cloning plasmids optimized to use with the Thermo Scientific 1-Step Human In Vitro Protein Expression System for &italic_begin;in vitro&italic_end; translation (IVT) of tagged fusion proteins.
The pT7CFE1 Expression Vectors contain the Encephalomyocarditis virus (EMCV) internal ribosome entry site (IRES) element that is critical for high levels of cap-independent protein expression in the Human In Vitro Translation System. Each vector features a highly-compatible multiple cloning site (MCS) to facilitate easy insertion of protein coding sequences into and between vectors. The pT7CFE1 Vector is available with single or tandem affinity tags at the N- or C- terminus to facilitate protein purification and detection. The pT7CFE Vectors are suitable for insertion of cloned genes, cDNAs, ORFs or PCR products for in vitro transcription and translation. Custom cloning services are also available.
Highlights:
  • Optimized performance – designed to provide the highest yield in the human in vitro translation system
  • Many options – multiple tag and tag-location options available
  • Modular MCS – multiple cloning site is maintained across vector family to facilitate subcloning
  • Cleavable tags – HRV 3C cleavage site available on select vectors
pT7CFE1 Plasmid Features:
  • EMCV IRES at the 5-prime UTR promotes high-level translation of mRNAs
  • MCS accommodates gene insertion via ten different restriction sites: Msc1, Nde1, BamH1, EcoR1, EcoRV, Pac1, Pst1, Sac1, Sal1, Not1 and Xho1
  • Poly A sequence in the 3-prime region promotes mRNA stabilization and protection from nucleases
  • T7 terminator ensures synthesis of accurate size mRNA transcripts
  • Plasmid linearization may be accomplished with restriction sites between Poly A sequence and the T7 terminator region