Please login to your online account to display your discounted pricing

PT 1, Tocris™

AMPK activator

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material PT 1
Assay Percent Range >98%
Molecular Formula C23H16ClN3O6S
Formula Weight 497.91
Synonym 2-Chloro-5-[[5-[[5-(4, 5-Dimethyl-2-nitrophenyl)-2-furanyl]methylene]-4, 5-dihydro-4-oxo-2-thiazolyl]amino]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

403910

 
r & d systems
RDS403910
10mg Each for $246.52

403950

 
r & d systems
RDS403950
50mg Each for $1,023.37
Description & Specifications

Specifications

Formula Weight 497.91
Quantity 10mg
Chemical Name or Material PT 1
Molecular Formula C23H16ClN3O6S
Assay Percent Range >98%
CAS 331002-70-1
Synonym 2-Chloro-5-[[5-[[5-(4, 5-Dimethyl-2-nitrophenyl)-2-furanyl]methylene]-4, 5-dihydro-4-oxo-2-thiazolyl]amino]benzoic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C