missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Psoralen, 97%

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

$320.76 - $1,287.68

Chemical Identifiers

CAS 66-97-7
Molecular Formula C11H6O3
Molecular Weight (g/mol) 186.166
MDL Number MFCD00010520
InChI Key ZCCUUQDIBDJBTK-UHFFFAOYSA-N
Synonym psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
PubChem CID 6199
ChEBI CHEBI:27616
IUPAC Name furo[3,2-g]chromen-7-one
SMILES C1=CC(=O)OC2=CC3=C(C=CO3)C=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60042MC View Documents Alfa Aesar™
J60042MC
100mg Each for $320.76
 
AAJ60042ME View Documents Alfa Aesar™
J60042ME
500mg Each for $1,287.68
 
Specifications

Chemical Identifiers

66-97-7
186.166
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
6199
furo[3,2-g]chromen-7-one
C11H6O3
MFCD00010520
psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
CHEBI:27616
C1=CC(=O)OC2=CC3=C(C=CO3)C=C21

Specifications

White
100mg
C11H6O3
152784
psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
C1=CC(=O)OC2=CC3=C(C=CO3)C=C21
186.166
CHEBI:27616
Solid
Psoralen
163°C to 164°C
66-97-7
MFCD00010520
14,7928
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
furo[3,2-g]chromen-7-one
6199
186.16
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LV0944000

Recommended Storage : 2° to 8°C