Please login to your online account to display your discounted pricing

Psora 4, Tocris™

Potent KV1.3 channel blocker

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Psora 4
Assay Percent Range >99%
Molecular Formula C21H18O4
Formula Weight 334.37
Synonym 4-(4-Phenylbutoxy)-7H-furo[3, 2-g][1]benzopyran-7-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

436710

 
r & d systems
RDS436710
10mg Each for $239.63

436750

 
r & d systems
RDS436750
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 334.37
Quantity 10mg
Chemical Name or Material Psora 4
Assay Percent Range >99%
Molecular Formula C21H18O4
CAS 724709-68-6
Synonym 4-(4-Phenylbutoxy)-7H-furo[3, 2-g][1]benzopyran-7-one
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C