Please login to your online account to display your discounted pricing

PSN 375963 hydrochloride, Tocris

GPR119 receptor agonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material PSN 375963 hydrochloride
Assay Percent Range >98%
Molecular Formula C17H23N3O·HCl
Formula Weight 321.84
Synonym 4-[5-(4-Butylcyclohexyl)-1, 2, 4-oxadiazol-3-yl]-pyridine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

335310

 
r & d systems
RDS335310
10mg Each for $244.81

335350

 
r & d systems
RDS335350
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 321.84
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C17H23N3O·HCl
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material PSN 375963 hydrochloride
CAS 388575-52-8
Synonym 4-[5-(4-Butylcyclohexyl)-1, 2, 4-oxadiazol-3-yl]-pyridine hydrochloride
Safety and Handling

Desiccate at Room Temperature