Please login to your online account to display your discounted pricing

PSB 36, Tocris

Potent and selective A1 antagonist

$218.74 - $911.19

Chemical Identifiers

Chemical Name or Material PSB 36
Assay Percent Range >99%
Molecular Formula C21H30N4O3
Formula Weight 386.49
Synonym 1-Butyl-8-(hexahydro-2,5-methanopentalen-3a(1H)-yl)-3,7-dihydro-3-(3-hydroxypropyl)-1H-purine-2,6-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

201910

 
r & d systems
201910
10mg Each for $218.74

201950

 
r & d systems
201950
50mg Each for $911.19
Description & Specifications

Specifications

Formula Weight 386.49
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C21H30N4O3
CAS 524944-72-7
Chemical Name or Material PSB 36
Synonym 1-Butyl-8-(hexahydro-2,5-methanopentalen-3a(1H)-yl)-3,7-dihydro-3-(3-hydroxypropyl)-1H-purine-2,6-dione
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C