Please login to your online account to display your discounted pricing

PSB 069, Tocris

Non-selective NTPDase inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material PSB 069
Assay Percent Range >98%
Molecular Formula C20H12N2O5ClSNa
Formula Weight 450.83
Synonym 1-Amino-4-(4-chlorophenyl)aminoanthraquinone-2-sulfonic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

257350

 
r & d systems
RDS257350
50mg Each for $844.73

257310

 
r & d systems
RDS257310
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 450.83
Quantity 50mg
Chemical Name or Material PSB 069
Molecular Formula C20H12N2O5ClSNa
CAS 78510-31-3
Synonym 1-Amino-4-(4-chlorophenyl)aminoanthraquinone-2-sulfonic acid sodium salt
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature