missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody.

$241.86 - $1,157.39

Specifications

Antigen pS2/pNR-2 Estrogen-Regulated Protein Ab-2
Clone GE2; previously R47/94
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS199P Thermo Scientific™ Lab Vision™
MS199P
Purified Ab with BSA and Azide 1mL Each for $1,059.06
MS199P0 Thermo Scientific™ Lab Vision™
MS199P0
Purified Ab with BSA and Azide 100μL Each for $241.86
MS199P1 Thermo Scientific™ Lab Vision™
MS199P1
Purified Ab with BSA and Azide 500μL Each for $706.02
MS199P1ABX Thermo Scientific™ Lab Vision™
MS199P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $800.12
MS199PABX Thermo Scientific™ Lab Vision™
MS199PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,157.39
MS199R7 Thermo Scientific™ Lab Vision™
MS199R7
Ready-to-use for IHC (UVLP) 7mL Each for $673.04
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody.

pS2 is a cysteine-rich, 6.5kDa protein found in both estrogen-dependent (breast tumors) and estrogen-independent tissues (normal stomach mucosa). About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is primarily expressed in estrogen receptor-positive breast tumors.

Host Species: Mouse

Clone: GE2; same as R47/94

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 57-84

Immunogen: A synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein

Molecular Weight: 6.5kDa

Positive Control: Normal stomach or breast CA

Cellular Localization: Cytoplasmic Antibody to pS2 is reportedly useful in identifying a subset of estrogen-dependent breast tumors which may respond to endocrine therapy.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

pS2/pNR-2 Estrogen-Regulated Protein Ab-2
Immunohistochemistry (Paraffin)
Unlabeled
Murine
Cancer and Tumor Biology
A Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein
GE2; previously R47/94
Monoclonal
Human
RUO
Primary
IgG1