missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS199P1ABX

 View more versions of this product

Catalog No. MS199P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody.

pS2 is a cysteine-rich, 6.5kDa protein found in both estrogen-dependent (breast tumors) and estrogen-independent tissues (normal stomach mucosa). About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is primarily expressed in estrogen receptor-positive breast tumors.

Host Species: Mouse

Clone: GE2; same as R47/94

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 57-84

Immunogen: A synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein

Molecular Weight: 6.5kDa

Positive Control: Normal stomach or breast CA

Cellular Localization: Cytoplasmic Antibody to pS2 is reportedly useful in identifying a subset of estrogen-dependent breast tumors which may respond to endocrine therapy.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

pS2/pNR-2 Estrogen-Regulated Protein Ab-2
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1
RUO
Primary
Human
Immunohistochemistry (Paraffin)
GE2; previously R47/94
Unlabeled
Concentrated
A Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein
100μL
Cancer and Tumor Biology
Monoclonal