missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PS 48, Tocris™

PDK1 activator

$138.08 - $581.48

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C17H15ClO2
Formula Weight 286.75
Synonym (2Z)-5-(4-Chlorophenyl)-3-phenyl-2-pentenoic acid
Chemical Name or Material PS 48
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
408750 R & D Systems
408750
50mg Each for $581.48
408710 R & D Systems
408710
10mg Each for $138.08
Specifications Description & Specifications

Chemical Identifiers

>98%
286.75
PS 48
C17H15ClO2
(2Z)-5-(4-Chlorophenyl)-3-phenyl-2-pentenoic acid

Specifications

50mg
>98%
286.75
PS 48
1180676-32-7
C17H15ClO2
(2Z)-5-(4-Chlorophenyl)-3-phenyl-2-pentenoic acid

Safety and Handling

Recommended Storage : Store at +4°C