missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Prulifloxacin, TCI America™

Manufacturer:  TCI America P20585G

 View more versions of this product

Catalog No. P20585G



Specifications

Specifications

Prulifloxacin
220°C
123447-62-1
MFCD00864847
CC1N2C3=CC(=C(C=C3C(=O)C(=C2S1)C(=O)O)F)N4CCN(CC4)CC5=C(OC(=O)O5)C
461.464
461.46
≥98.0% (HPLC,T)
Yellow
5g
C21H20FN3O6S
PWNMXPDKBYZCOO-UHFFFAOYSA-N
6-fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]piperazin-1-yl]-4-oxo-1H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
65947
Crystal-Powder at 20°C

Chemical Identifiers

123447-62-1
461.464
PWNMXPDKBYZCOO-UHFFFAOYSA-N
6-fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]piperazin-1-yl]-4-oxo-1H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
C21H20FN3O6S
MFCD00864847
65947
CC1N2C3=CC(=C(C=C3C(=O)C(=C2S1)C(=O)O)F)N4CCN(CC4)CC5=C(OC(=O)O5)C
Safety and Handling

Safety and Handling

RTECSNumber : XJ0600000

TSCA : No

Recommended Storage : Freezer

Documents