missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ PRT 4165

Inhibitor of Bmi1/Ring1A; blocks histone H2A ubiquitination

$97.76 - $410.62

Specifications

Product Type PRT 4165
Inhibitors PRT 4165
Molecular Weight 235.24
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
504710R Tocris™
RDS504710
10mg Each for $97.76
504750R Tocris™
RDS504750
50mg Each for $410.62
Specifications

Description

Inhibitor of Bmi1/Ring1A, subunits of the polycomb repressive complex 1 (PRC1); inhibits self-ubiquitination (IC50 = 3.9μM) but does not increase cellular levels of either subunit. Prevents Bmi1/Ring1A-mediated ubiquitination and drug-induced degradation of topoisomerase 2α (Top2α). Also shown to inhibit the in vitro E3 ubiquitin ligase activity of RNF2 and a Bmi1/RNF2 complex, inhibiting H2A/H2AX ubiquitination. Blocks polycomb repressor complex (PRC) 1-mediated histone H2A ubiquitination in vitro and in vivo.
  • Chemical Name: 2-(3-Pyridinylmethylene)-1H-Indene-1,3(2H)-dione
  • Formula: C15H9NO2
  • CAS Number: 31083-55-3
  • Solubility: Soluble to 100mM in DMSO and to 5mM in 1eq. HCl with gentle warming
  • Purity: >99
Description & Specifications

Specifications

PRT 4165
PRT 4165
235.24
>99