Please login to your online account to display your discounted pricing

Proxyfan oxalate, Tocris

High affinity H3 ligand

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Proxyfan oxalate
Assay Percent Range >98%
Molecular Formula C13H16N2O·C2H2O4
Formula Weight 306.32
Synonym 4-[3-(Phenylmethoxy)propyl]-1H-imidazole oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

247750

 
r & d systems
RDS247750
50mg Each for $746.48

247710

 
r & d systems
RDS247710
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 306.32
Quantity 50mg
Chemical Name or Material Proxyfan oxalate
Molecular Formula C13H16N2O·C2H2O4
Assay Percent Range >98%
CAS 177708-09-7
Synonym 4-[3-(Phenylmethoxy)propyl]-1H-imidazole oxalate
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature