Please login to your online account to display your discounted pricing

Protocatechuic acid, 99.9%, For HPLC analysis, MP Biomedicals™


Quantity 10g

Manufacturer: mp biomedicals  0215642110

 View more versions of this product

Catalog No. ICN15642110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10g
Chemical Name or Material Protocatechuic acid
Name Note For HPLC analysis
CAS 99-50-3
Physical Form Crystalline
Assay Percent Range 0.999
Molecular Formula C7H6O4
Formula Weight 154.1g/mol
Synonym 3, 4-Dihydroxybenzoic acid
Recommended Storage Store at Room Temperature.
EINECS Number 202-760-0

Specifications

Chemical Name or Material Protocatechuic acid
Name Note For HPLC analysis
CAS 99-50-3
Physical Form Crystalline
Assay Percent Range 0.999
Molecular Formula C7H6O4
Formula Weight 154.1g/mol
Synonym 3, 4-Dihydroxybenzoic acid
Accessories

Store at Room Temperature.