Please login to your online account to display your discounted pricing

Protocatechuic acid, 99.9%, For HPLC analysis, MP Biomedicals™

$99.53 - $99.53

Chemical Identifiers

Chemical Name or Material Protocatechuic acid
Name Note For HPLC analysis
CAS 99-50-3
Physical Form Crystalline
Assay Percent Range 0.999
Molecular Formula C7H6O4
Formula Weight 154.1g/mol
Synonym 3, 4-Dihydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15642110

 
mp biomedicals
0215642110
10g Each for $99.53
Description & Specifications

Specifications

Quantity 10g
Chemical Name or Material Protocatechuic acid
Name Note For HPLC analysis
CAS 99-50-3
Physical Form Crystalline
Assay Percent Range 0.999
Molecular Formula C7H6O4
Formula Weight 154.1g/mol
Synonym 3, 4-Dihydroxybenzoic acid
Recommended Storage Store at Room Temperature.
EINECS Number 202-760-0
Safety and Handling

Store at Room Temperature.