missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Prothionamide, TCI America™

Manufacturer:  TCI America P230225G

 View more versions of this product

Catalog No. P230225G



Specifications

Specifications

Prothionamide
≥98.0% (HPLC,T)
MFCD00464119
Yellow
2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide
CCCC1=NC=CC(=C1)C(=S)N
180.269
180.27
14222-60-7
C9H12N2S
143°C
25g
VRDIULHPQTYCLN-UHFFFAOYSA-N
2-propylpyridine-4-carbothioamide
666418
Crystal-Powder at 20°C

Chemical Identifiers

14222-60-7
180.269
VRDIULHPQTYCLN-UHFFFAOYSA-N
666418
CCCC1=NC=CC(=C1)C(=S)N
C9H12N2S
MFCD00464119
2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide
2-propylpyridine-4-carbothioamide
Safety and Handling

Safety and Handling

RTECSNumber : NS0650000

TSCA : No

Documents