Please login to your online account to display your discounted pricing

Alfa Aesar™ Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control

For cellular and molecular biology applications

Specifications

Product Type Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Molecular Weight 843.98
Storage Requirements -30° to -10°C
Sequence Leu-Ser-Glu-Thr-Lys-Pro-Ala-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66521EXD

 
alfa aesar™
J66521EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66521MCR

 
alfa aesar™
J66521MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Molecular Weight 843.98
Storage Requirements -30° to -10°C
Sequence Leu-Ser-Glu-Thr-Lys-Pro-Ala-Val
Format Solid
Regulatory Status RUO
Purification Method HPLC