missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Protein Kinase C-epsilon Peptide Substrate

For cellular and molecular biology applications

Specifications

Storage Requirements -30° to -10°C
Color White
Molecular Weight 2067.45
Format Lyophilized
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
AAJ66938EXD Alfa Aesar™
J66938EXD
0.5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66938MCR Alfa Aesar™
J66938MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

-30° to -10°C
White
2067.45
Lyophilized