Please login to your online account to display your discounted pricing

Alfa Aesar™ Protein Kinase C (19-36)

For cellular and molecular biology applications

Specifications

Product Type Protein Kinase C (19-36)
Molecular Weight 2151.51
Storage Requirements -30° to -10°C
Sequence Arg-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln-Lys-Asn-Val-His-Glu-Val-Lys-Asn
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66426EXD

 
alfa aesar™
J66426EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66426MCR

 
alfa aesar™
J66426MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Protein Kinase C (19-36)
Molecular Weight 2151.51
Storage Requirements -30° to -10°C
Sequence Arg-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln-Lys-Asn-Val-His-Glu-Val-Lys-Asn
Format Solid
Common Name PKC Inhibitor Peptide
Regulatory Status RUO
Purification Method HPLC