Please login to your online account to display your discounted pricing

Protein Disulfide Isomerase/P4HB Mouse anti-Human, Clone: 537331, R&D Systems

Mouse Monoclonal Antibody

$317.73 - $317.73

Specifications

Antigen Protein Disulfide Isomerase/P4HB
Clone 537331
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Protein Disulfide Isomerase/P4HB; AA Info: Asp18-Lys505
Gene ID (Entrez) 5034
Gene Accession No. P07237
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

MAB4236

 
r & d systems
RDSMAB4236
100μg Each for $317.73
Description & Specifications

Specifications

Antigen Protein Disulfide Isomerase/P4HB
Clone 537331
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Protein Disulfide Isomerase/P4HB; AA Info: Asp18-Lys505
Gene ID (Entrez) 5034
Gene Accession No. P07237
Formulation Supplied as a 0.2μm filtered solution in PBS.
Reconstitution Sterile PBS to a final concentration of 0.5mg/mL.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Mouse
Cross Reactivity Human
Test Specificity Detects human Protein Disulfide Isomerase/P4HB in direct ELISAs and Western blots.
Isotype IgG2A
Conjugate Unlabeled
Purification Method Protein A or G purified from hybridoma culture supernatant
Format Purified
Applications Immunocytochemistry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Storage Requirements -20 to -70°C as supplied; 1 month

Protein Disulfide Isomerase/P4HB Monoclonal antibody MAB4236 Specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting