Please login to your online account to display your discounted pricing
Labconco™ Protector™ ClassMate™ SpillStopper™ Work Surfaces
Dished and contoured to conform to interior liner of Protector Classmate Laboratory Hoods.
$2,045.25 - $2,308.50
Specifications
Depth (Metric) 66cm
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Includes Weight (Metric) Width (Metric) Price Quantity    
S15393
labconco™
6986400EMD
4 ft. Protector ClassMate Laboratory Hood No cupsink cutout 50kg 122cm Each for $2,045.25
S15395
labconco™
6986402
4 ft. Protector ClassMate Laboratory Hood Right rear cupsink cutout 50kg 122cm Each for $2,308.50
Description & Specifications
Specifications
Depth (Metric) 66cm
  • Molded from corrosion-resistant epoxy resins
  • Contoured to conform to interior liner
  • Dished design helps to contain spills
  • Front edge has large radius to direct airflow into the hood
  • Pre-drilled holes for venting and plumbing passthrough
  • May be ordered with oval cupsink cutout(s)
Cupsink sold separately.