missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Proteasome Substrate

For cellular and molecular biology applications

Specifications

Substrate Type Fluorogenic
Storage Requirements -30° to -10°C
For Use With (Application) Fluorogenic proteasome substrate
Color White
Molecular Weight 648.79
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
AAJ64492LB0 Alfa Aesar™
J64492LB0
5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ64492MCR Alfa Aesar™
J64492MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

Fluorogenic
Fluorogenic proteasome substrate
648.79
-30° to -10°C
White
Lyophilized