missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Protamine sulfate

$55.05 - $262.16

Chemical Identifiers

CAS 9009-65-8
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132091
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6292603 View Documents Alfa Aesar™
J6292603
1g Each for $55.05
 
AAJ6292606 View Documents Alfa Aesar™
J6292606
5g Each for $150.81
 
AAJ6292609 View Documents Alfa Aesar™
J6292609
10g Each for $262.16
 
Description

Description

Solubility
It is sparingly soluble in water.

Notes
Hygroscopic. Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

9009-65-8
0.00
MFCD00132091

Specifications

Hygroscopic
1g
MFCD00132091
Powder
White
9009-65-8
0.00
Protamine sulfate
Safety and Handling

Safety and Handling

RTECSNumber : UK9450000

TSCA : TSCA