Please login to your online account to display your discounted pricing
ProSwift™ C4 RP-5H Capillary LC Columns
Improve speed and resolution of LC-MS protein identification and biomarker discovery using these C4 monolithic HPLC columns.
$1,712.32 - $2,311.65
Column Chemistry Butyl (C4)
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Column Type Diameter (Metric) Length (Metric) Particle Size Price Quantity    
DX164928
thermo fisher scientific
164928
Reversed Phase 0.1mm 500mm Monolith Each for $2,311.65
DX164929
thermo fisher scientific
164929
Reversed Phase 0.1mm 250mm Monolith Each for $2,140.42
DX164930
thermo fisher scientific
164930
Reversed Phase 0.2mm 250mm Monolith Each for $2,140.42
DX164931
thermo fisher scientific
164931
Reversed Phase 0.5mm 100mm Monolith Each for $1,712.32
DX164932
thermo fisher scientific
164932
Reversed Phase 0.5mm 250mm Monolith Each for $2,140.42
DX164935
thermo fisher scientific
164935
Reversed Phase 0.05mm 250mm Monolith Each for $2,311.65
Description & Specifications
Specifications
Column Chemistry Butyl (C4)
Column Type Reversed Phase
Particle Size Monolith
For Use With LC/MS
Material Poly(ethylenedimethacrylate-co-butylmethacrylate)
Packing Material Monolith
Pore Size 1.5µm
Solvents Most Common Organic Solvents
Stationary Phase Butyl (C4)
Temperature 90°C
Type Analytical Column
USP Type NA
Improve speed and resolution of protein identification and biomarker discovery by LC-MS. Thermo Scientific™ ProSwift™ C4 Monolithic HPLC columns provide fast, high-resolution LC-MS separation for protein identification, biomarker discovery, and systems biology applications. Based on a cross-linked butyl methacrylate co-polymer, the monolithic structure offers a high-quality alternative to traditional micro-particulate sorbents, providing important advantages to the chromatographic separation. 
Easy High-Sensitivity Proteomics and Biotech Applications
Each monolith is a single uninterrupted, interconnected network of flow-through channels of a specific pore size. These channels and the monolith's nonporous surfaces provide fast mass transfer for high-resolution and fast protein separations. The monolith morphology results in low operating backpressure, allowing the use of higher linear velocities which can improve resolution of large biomolecules. 
Column Features 
• Low carryover
• High-speed protein separations
• High column-to-column reproducibility
• Wide range of column I.D. and lengths available
• Superior lifetime
• Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings