missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Prostaglandin E2 Parameter Assay Kit, R&D Systems

a 3.5 hour forward sequential competitive enzyme immunoassay designed to measure PGE2 in cell culture supernates, serum, plasma, and urine

$415.83 - $1,995.75

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
For Use With (Application) ELISA
Sensitivity 414pg/mL
Species Multi-species
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Sufficient For Product Type Quantity Price Quantity  
KGE004B R & D Systems
RDSKGE004B
1 x 96-well plate ELISA kit 1 kit Each for $415.83
SKGE004B R & D Systems
RDSSKGE004B
6 x 96-well plates ELISA kit 1 kit Each for $1,995.75
Specifications

Description

Assay Type: Solid Phase Competitive ELISA
Sample Type/Volume Required: Cell Culture Supernates (51μL), Serum (51μL), EDTA Plasma (51μL), Heparin Plasma (51μL), Urine (51μL)
Sensitivity: 41.4pg/mL
Specificity: Measures PGE2 levels in various samples
Cross Reactivity: Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
ELISA
414pg/mL
Multi-species