Please login to your online account to display your discounted pricing

Prostaglandin E2 Parameter Assay Kit, R&D Systems


Manufacturer: r & d systems  RDSSKGE004B

 View more versions of this product

Catalog No. SKGE004B

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Quantity 1 kit
For Use With (Application) For the quantitative determination of Prostaglandin E2 (PGE2) in cell culture supernates, serum, plasma and urine
Sensitivity 414pg/mL
Product Type ELISA kit
Species Multi-species
Sufficient For 6 x 96-well plates