Please login to your online account to display your discounted pricing

Alfa Aesar™ Prostaglandin B2, 99%

Chemical Identifiers

Chemical Name or Material Prostaglandin B2
CAS 13367-85-6
Formula Weight 334.46
MDL Number MFCD00065730
Synonym PGB2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67293MCR

 
alfa aesar™
J67293MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 334.46
Chemical Name or Material Prostaglandin B2
MDL Number MFCD00065730
Physical Form Waxy Solid
Assay 99%
Synonym PGB2
CAS 13367-85-6
Storage Note 1 -30° to -10°C
Color White to off-white
Quantity 1mg
Safety and Handling

-30° to -10°C